missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Chloro-4-nitrophenyl)homopiperazine, 96%

Manufacturer:  Alfa Aesar™ H6310506

 View more versions of this product

Catalog No. AAH6310506



Specifications

Specifications

1-(2-Chloro-4-nitrophenyl)homopiperazine
220220-19-9
UN3259
MVZOFBIWSFBCKA-UHFFFAOYSA-N
1-(2-chloro-4-nitrophenyl)-1,4-diazepane
540495
Solid
5g
C11H14ClN3O2
1-2-chloro-4-nitrophenyl-1,4-diazepane, 1-2-chloro-4-nitro-phenyl-1,4 diazepane, 1h-1,4-diazepine, 1-2-chloro-4-nitrophenyl hexahydro, 1-2-chloro-4-nitrophenyl-1,4-diazaperhydroepine
C1CNCCN(C1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
255.702
255.7
96%

Chemical Identifiers

220220-19-9
255.702
1-2-chloro-4-nitrophenyl-1,4-diazepane, 1-2-chloro-4-nitro-phenyl-1,4 diazepane, 1h-1,4-diazepine, 1-2-chloro-4-nitrophenyl hexahydro, 1-2-chloro-4-nitrophenyl-1,4-diazaperhydroepine
1-(2-chloro-4-nitrophenyl)-1,4-diazepane
C11H14ClN3O2
MVZOFBIWSFBCKA-UHFFFAOYSA-N
540495
C1CNCCN(C1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

SDS Documents