Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Chloro-4-nitrophenyl)-4-piperidone, 97%

Chemical Identifiers

Chemical Name or Material 1-(2-Chloro-4-nitrophenyl)-4-piperidone
Formula Weight 254.67
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6386503

 
alfa aesar™
H6386503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6386506

 
alfa aesar™
H6386506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63865MD

 
alfa aesar™
H63865MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 254.67
Chemical Name or Material 1-(2-Chloro-4-nitrophenyl)-4-piperidone
Physical Form Solid
Assay 97%
Quantity 1g