Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Chloro-4-nitrophenyl)-4-methylpiperazine, 97%

Chemical Identifiers

Chemical Name or Material 1-(2-Chloro-4-nitrophenyl)-4-methylpiperazine
CAS 16154-62-4
Formula Weight 255.7
MDL Number MFCD00030787
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6339703

 
alfa aesar™
H6339703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6339706

 
alfa aesar™
H6339706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63397MD

 
alfa aesar™
H63397MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 255.7
Chemical Name or Material 1-(2-Chloro-4-nitrophenyl)-4-methylpiperazine
Physical Form Solid
Assay 97%
MDL Number MFCD00030787
CAS 16154-62-4
Quantity 1g