Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Chloro-4-nitrophenyl)-4-ethylpiperazine, 97%

Chemical Identifiers

Chemical Name or Material 1-(2-Chloro-4-nitrophenyl)-4-ethylpiperazine
Formula Weight 269.73
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6377303

 
alfa aesar™
H6377303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6377306

 
alfa aesar™
H6377306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63773MD

 
alfa aesar™
H63773MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 269.73
Chemical Name or Material 1-(2-Chloro-4-nitrophenyl)-4-ethylpiperazine
Physical Form Solid
Assay 97%
Quantity 1g