missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2-Butylene Carbonate 98.0+%, TCI America™
Manufacturer: Tci America B332125G
Specifications
1,2-Butylene Carbonate | |
Yellow | |
4437-85-8 | |
MFCD00143317 | |
ZZXUZKXVROWEIF-UHFFFAOYSA-N | |
4-ethyl-1,3-dioxolan-2-one | |
107282 | |
Clear Liquid at 20°C |
74°C | |
25g | |
C5H8O3 | |
1,2-butanediol carbonate, 1,2-butanediol, cyclic carbonate, 1,2-butylene carbonate, 1,3-dioxolan-2-one, 4-ethyl, 1,3-dioxolan-2-one, 4-ethyl-, s, acmc-1ak00, acmc-20ltqj, butylene carbonate, carbonic acid, cyclic ethylethylene ester, dsstox_cid_20342 | |
CCC1COC(=O)O1 | |
116.116 | |
116.12 | |
≥98.0% (GC) |
Chemical Identifiers
4437-85-8 | |
116.116 | |
ZZXUZKXVROWEIF-UHFFFAOYSA-N | |
107282 | |
CCC1COC(=O)O1 |
C5H8O3 | |
MFCD00143317 | |
1,2-butanediol carbonate, 1,2-butanediol, cyclic carbonate, 1,2-butylene carbonate, 1,3-dioxolan-2-one, 4-ethyl, 1,3-dioxolan-2-one, 4-ethyl-, s, acmc-1ak00, acmc-20ltqj, butylene carbonate, carbonic acid, cyclic ethylethylene ester, dsstox_cid_20342 | |
4-ethyl-1,3-dioxolan-2-one |
Safety and Handling
EINECSNumber : (5)-6238
RTECSNumber : JI4582000
TSCA : Yes