missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Bromophenylsulfonyl)-3-methylpiperidine, 97%

Chemical Identifiers

CAS 1184609-55-9
Molecular Formula C12H16BrNO2S
Molecular Weight (g/mol) 318.229
MDL Number MFCD12650340
InChI Key DHHOXSFLPKIJME-UHFFFAOYSA-N
Synonym 1-2-bromobenzenesulfonyl-3-methylpiperidine, 1-2-bromophenylsulfonyl-3-methylpiperidine
PubChem CID 51169806
IUPAC Name 1-(2-bromophenyl)sulfonyl-3-methylpiperidine
SMILES CC1CCCN(C1)S(=O)(=O)C2=CC=CC=C2Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60206MD
View Documents
Alfa Aesar™
H60206MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6020603
View Documents
Alfa Aesar™
H6020603
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Applications
1-(2-Bromophenylsulfonyl)-3-methylpiperidine is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

1184609-55-9
318.229
DHHOXSFLPKIJME-UHFFFAOYSA-N
51169806
CC1CCCN(C1)S(=O)(=O)C2=CC=CC=C2Br
C12H16BrNO2S
MFCD12650340
1-2-bromobenzenesulfonyl-3-methylpiperidine, 1-2-bromophenylsulfonyl-3-methylpiperidine
1-(2-bromophenyl)sulfonyl-3-methylpiperidine

Specifications

250mg
C12H16BrNO2S
1-2-bromobenzenesulfonyl-3-methylpiperidine, 1-2-bromophenylsulfonyl-3-methylpiperidine
CC1CCCN(C1)S(=O)(=O)C2=CC=CC=C2Br
318.229
318.23
1-(2-Bromophenylsulfonyl)-3-methylpiperidine
1184609-55-9
MFCD12650340
DHHOXSFLPKIJME-UHFFFAOYSA-N
1-(2-bromophenyl)sulfonyl-3-methylpiperidine
51169806
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures