missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Bromophenylsulfonyl)-3-methylpiperidine, 97%

Manufacturer:  Alfa Aesar™ H6020603

 View more versions of this product

Catalog No. AAH6020603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Applications
1-(2-Bromophenylsulfonyl)-3-methylpiperidine is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

1-(2-Bromophenylsulfonyl)-3-methylpiperidine
1184609-55-9
MFCD12650340
DHHOXSFLPKIJME-UHFFFAOYSA-N
1-(2-bromophenyl)sulfonyl-3-methylpiperidine
51169806
97%
1g
C12H16BrNO2S
1-2-bromobenzenesulfonyl-3-methylpiperidine, 1-2-bromophenylsulfonyl-3-methylpiperidine
CC1CCCN(C1)S(=O)(=O)C2=CC=CC=C2Br
318.229
318.23

Chemical Identifiers

1184609-55-9
318.229
DHHOXSFLPKIJME-UHFFFAOYSA-N
51169806
CC1CCCN(C1)S(=O)(=O)C2=CC=CC=C2Br
C12H16BrNO2S
MFCD12650340
1-2-bromobenzenesulfonyl-3-methylpiperidine, 1-2-bromophenylsulfonyl-3-methylpiperidine
1-(2-bromophenyl)sulfonyl-3-methylpiperidine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents