missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Bromoethyl)pyrrole, TCI America™

$236.81 - $792.94

Chemical Identifiers

CAS 78358-86-8
Molecular Formula C6H8BrN
Molecular Weight (g/mol) 174.041
MDL Number MFCD00191322
InChI Key QBAVHEZVBGASER-UHFFFAOYSA-N
PubChem CID 11651225
IUPAC Name 1-(2-bromoethyl)pyrrole
SMILES C1=CN(C=C1)CCBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B16221G View Documents TCI America
B16221G
1g Each for $236.81
 
B16225G View Documents TCI America
B16225G
5g Each for $792.94
 
Specifications

Chemical Identifiers

78358-86-8
174.041
QBAVHEZVBGASER-UHFFFAOYSA-N
1-(2-bromoethyl)pyrrole
C6H8BrN
MFCD00191322
11651225
C1=CN(C=C1)CCBr

Specifications

Yellow
78358-86-8
MFCD00191322
C1=CN(C=C1)CCBr
174.041
174.04
≥97.0% (GC)
1g
C6H8BrN
QBAVHEZVBGASER-UHFFFAOYSA-N
1-(2-bromoethyl)pyrrole
11651225
Clear Liquid at 20°C
1-(2-Bromoethyl)pyrrole
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator