missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Bromoethyl)naphthalene, 97%

Manufacturer:  Alfa Aesar™ 4349114

 View more versions of this product

Catalog No. AA4349114



Specifications

Specifications

1- (2-Bromoethyl)naphthalene
237°C to 238°C (1.65mmHg)
13686-49-2
MFCD00037737
GPHCPUFIWQJZOI-UHFFFAOYSA-N
1-(2-bromoethyl)naphthalene
139541
Liquid
>110°C (230°F)
25g
C12H11Br
1-2-bromo-ethyl-naphthalene
C1=CC=C2C(=C1)C=CC=C2CCBr
235.124
235.13
97%

Chemical Identifiers

13686-49-2
235.124
GPHCPUFIWQJZOI-UHFFFAOYSA-N
139541
C1=CC=C2C(=C1)C=CC=C2CCBr
C12H11Br
MFCD00037737
1-2-bromo-ethyl-naphthalene
1-(2-bromoethyl)naphthalene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents