Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Bromoethyl)-4-nitrobenzene, 98%

$71.72 - $227.81

Chemical Identifiers

Chemical Name or Material 1-(2-Bromoethyl)-4-nitrobenzene
CAS 5339-26-4
Molecular Formula C8H8BrNO2
Formula Weight 230.07
MDL Number MFCD00007386
Beilstein 1909321
Synonym 4-Nitrophenethyl bromide; 2-(4-Nitrophenyl)ethyl bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1567706

  SDS
alfa aesar™
A1567706
5g Each for $71.72

AAA1567714

  SDS
alfa aesar™
A1567714
25g Each for $227.81
Description & Specifications

Specifications

Formula Weight 230.07
Chemical Name or Material 1-(2-Bromoethyl)-4-nitrobenzene
Assay 98%
Molecular Formula C8H8BrNO2
MDL Number MFCD00007386
CAS 5339-26-4
Melting Point 67° to 71°C
Beilstein 1909321
Synonym 4-Nitrophenethyl bromide; 2-(4-Nitrophenyl)ethyl bromide
Boiling Point 116°C (0.2mmHg)
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
1-(2-Bromoethyl)-4-nitrobenzene is an electrophile used as a substrate and activator for theta class gutathione transferase (GSTT-1). It is also used as a marker in for the θ-class of glutathione S-transferases (GSTs).

Solubility
Soluble in water (partly) and methanol.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away form oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.