Learn More
1-(2-Bromoethyl)-4-nitrobenzene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1567706
Description
1-(2-Bromoethyl)-4-nitrobenzene is an electrophile used as a substrate and activator for theta class gutathione transferase (GSTT-1). It is also used as a marker in for the θ-class of glutathione S-transferases (GSTs).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-(2-Bromoethyl)-4-nitrobenzene is an electrophile used as a substrate and activator for theta class gutathione transferase (GSTT-1). It is also used as a marker in for the θ-class of glutathione S-transferases (GSTs).
Solubility
Soluble in water (partly) and methanol.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away form oxidizing agent.
Specifications
1-(2-Bromoethyl)-4-nitrobenzene | |
67°C to 71°C | |
C8H8BrNO2 | |
5 g | |
4-nitrophenethyl bromide, 1-2-bromoethyl-4-nitrobenzene, benzene, 1-2-bromoethyl-4-nitro, 4-nitrophenethylbromide, p-nitrophenethyl bromide, 2-4-nitrophenyl ethyl bromide, 4-2-bromoethyl nitrobenzene, 4-nitrophenylethyl bromide, .beta.-p-nitrophenyl ethyl bromide, phenethylbromide4nitro | |
NTURQZFFJDCTMZ-UHFFFAOYSA-N | |
1-(2-bromoethyl)-4-nitrobenzene | |
79266 | |
98% |
5339-26-4 | |
116°C (0.2 mmHg) | |
MFCD00007386 | |
1909321 | |
Soluble in water (partly) and methanol. | |
C1=CC(=CC=C1CCBr)[N+](=O)[O-] | |
230.061 | |
230.07 |
Chemical Identifiers
5339-26-4 | |
230.061 | |
NTURQZFFJDCTMZ-UHFFFAOYSA-N | |
79266 | |
C1=CC(=CC=C1CCBr)[N+](=O)[O-] |
C8H8BrNO2 | |
MFCD00007386 | |
4-nitrophenethyl bromide, 1-2-bromoethyl-4-nitrobenzene, benzene, 1-2-bromoethyl-4-nitro, 4-nitrophenethylbromide, p-nitrophenethyl bromide, 2-4-nitrophenyl ethyl bromide, 4-2-bromoethyl nitrobenzene, 4-nitrophenylethyl bromide, .beta.-p-nitrophenyl ethyl bromide, phenethylbromide4nitro | |
1-(2-bromoethyl)-4-nitrobenzene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
H315-H317-H319-H335
EINECSNumber : 226-271-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only