missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Bromoethyl)-4-nitrobenzene, 98%

Manufacturer:  Alfa Aesar™ A1567714

 View more versions of this product

Catalog No. AAA1567714



Description

Description

Applications
1-(2-Bromoethyl)-4-nitrobenzene is an electrophile used as a substrate and activator for theta class gutathione transferase (GSTT-1). It is also used as a marker in for the θ-class of glutathione S-transferases (GSTs).

Solubility
Soluble in water (partly) and methanol.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away form oxidizing agent.
Specifications

Specifications

1-(2-Bromoethyl)-4-nitrobenzene
67°C to 71°C
5339-26-4
MFCD00007386
4-nitrophenethyl bromide, 1-2-bromoethyl-4-nitrobenzene, benzene, 1-2-bromoethyl-4-nitro, 4-nitrophenethylbromide, p-nitrophenethyl bromide, 2-4-nitrophenyl ethyl bromide, 4-2-bromoethyl nitrobenzene, 4-nitrophenylethyl bromide, .beta.-p-nitrophenyl ethyl bromide, phenethylbromide4nitro
C1=CC(=CC=C1CCBr)[N+](=O)[O-]
230.061
230.07
116°C (0.2mmHg)
25g
C8H8BrNO2
1909321
NTURQZFFJDCTMZ-UHFFFAOYSA-N
1-(2-bromoethyl)-4-nitrobenzene
79266
98%

Chemical Identifiers

5339-26-4
230.061
NTURQZFFJDCTMZ-UHFFFAOYSA-N
79266
C1=CC(=CC=C1CCBr)[N+](=O)[O-]
C8H8BrNO2
MFCD00007386
4-nitrophenethyl bromide, 1-2-bromoethyl-4-nitrobenzene, benzene, 1-2-bromoethyl-4-nitro, 4-nitrophenethylbromide, p-nitrophenethyl bromide, 2-4-nitrophenyl ethyl bromide, 4-2-bromoethyl nitrobenzene, 4-nitrophenylethyl bromide, .beta.-p-nitrophenyl ethyl bromide, phenethylbromide4nitro
1-(2-bromoethyl)-4-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents