Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Bromoethyl)-4-ethyl-1,4-dihydro-5H-tetrazol-5-one, 95%

Chemical Identifiers

Chemical Name or Material 1-(2-Bromoethyl)-4-ethyl-1,4-dihydro-5H-tetrazol-5-one
CAS 84501-67-7
Formula Weight 221.06
MDL Number MFCD16251259
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3652803

  SDS
alfa aesar™
H3652803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3652806

  SDS
alfa aesar™
H3652806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 221.06
Color Colorless
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
MDL Number MFCD16251259
CAS 84501-67-7
Physical Form Liquid
Refractive Index 1.5070
Density 1.777g/mL
Boiling Point 223°C
Assay 95%
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Chemical Name or Material 1-(2-Bromoethyl)-4-ethyl-1,4-dihydro-5H-tetrazol-5-one
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.