missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Bromoethoxy-4-bromobenzene) 98%, ACROS Organics™

Manufacturer:  Acros Organics AC147640050

 View more versions of this product

Catalog No. AC147640050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-(2-Bromoethoxy-4-bromobenzene)
Authentic
163.0°C to 165.0°C (16.0mmHg)
56.0°C to 57.0°C
18800-30-1
100.0
C8H8Br2O
MFCD00044201
1-bromo-4-2-bromoethoxy benzene, 2-bromoethyl p-bromophenyl ether, 4-bromo-1-2-bromoethoxy benzene, 2-bromoethyl 4-bromophenyl ether, benzene,1-bromo-4-2-bromoethoxy, benzene, 1-bromo-4-2-bromoethoxy, 4-bromophenoxyethyl bromide, phenetole, p,.beta.-dibromo, 2-4-bromophenoxy ethylbromide, 2-p-bromophenoxy ethyl bromide
C1=CC(=CC=C1OCCBr)Br
279.959
279.96
98%
180°C
Glass bottle
White to Beige
5g
97.5
98%
BrC6H4OCH2CH2Br
06, I, 105
SRGBSEMMGLSUMI-UHFFFAOYSA-N
1-bromo-4-(2-bromoethoxy)benzene
87797
Crystalline Powder or Crystals

Chemical Identifiers

18800-30-1
279.959
SRGBSEMMGLSUMI-UHFFFAOYSA-N
87797
C1=CC(=CC=C1OCCBr)Br
C8H8Br2O
MFCD00044201
1-bromo-4-2-bromoethoxy benzene, 2-bromoethyl p-bromophenyl ether, 4-bromo-1-2-bromoethoxy benzene, 2-bromoethyl 4-bromophenyl ether, benzene,1-bromo-4-2-bromoethoxy, benzene, 1-bromo-4-2-bromoethoxy, 4-bromophenoxyethyl bromide, phenetole, p,.beta.-dibromo, 2-4-bromophenoxy ethylbromide, 2-p-bromophenoxy ethyl bromide
1-bromo-4-(2-bromoethoxy)benzene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents