Please login to your online account to display your discounted pricing

1-(2-Bromoethoxy-4-bromobenzene) 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-(2-Bromoethoxy-4-bromobenzene)
CAS 18800-30-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H8Br2O
Linear Formula BrC6H4OCH2CH2Br
Formula Weight 279.96
MDL Number MFCD00044201
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC147640050

  SDS
fisher scientific acros
AC147640050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC147640250

  SDS
thermo fisher scientific
AC147640250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder or Crystals
Formula Weight 279.96
Quantity 5g
Color White to beige
Beilstein 06, I, 105
CAS Max % 100.0
Linear Formula BrC6H4OCH2CH2Br
Chemical Name or Material 1-(2-Bromoethoxy-4-bromobenzene)
CAS 18800-30-1
Assay Percent Range 98%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C8H8Br2O
MDL Number MFCD00044201
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.