Please login to your online account to display your discounted pricing

1-(2-Bromoethoxy)-2-bromobenzene, 95%, ACROS Organics™

$58.17 - $58.17

Chemical Identifiers

Chemical Name or Material 1-(2-Bromoethoxy)-2-bromobenzene
CAS 18800-28-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8H8Br2O
Linear Formula BrC6H4O(CH2)2Br
Formula Weight 279.96
MDL Number MFCD00634301
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC379220250

  SDS
thermo fisher scientific
AC379220250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC379220050

  SDS
thermo fisher scientific
AC379220050
Glass bottle 5g Each for $58.17
Description & Specifications

Specifications

Assay ≥94%
Physical Form Liquid After Melting
Formula Weight 279.96
Quantity 25g
Color Clear colorless to yellow
CAS Min % 94.0
CAS Max % 100.0
Packaging Glass bottle
Storage Information Product may darken during storage
Linear Formula BrC6H4O(CH2)2Br
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Infrared Spectrum Authentic
Chemical Name or Material 1-(2-Bromoethoxy)-2-bromobenzene
Assay Percent Range 95%
MDL Number MFCD00634301
CAS 18800-28-7
Health Hazard 1 GHS Signal Word: Warning
Molecular Formula C8H8Br2O
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.