missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-[2-(Boc-amino)ethyl]indole-6-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H6362706

 View more versions of this product

Catalog No. AAH6362706



Specifications

Specifications

1-[2-(Boc-amino)ethyl]indole-6-carboxylic acid
5g
C9H5N3O2S
4-nitro-1h-indol-3-yl sulfanyl formonitrile
C1=CC2=C(C(=C1)[N+](=O)[O-])C(=CN2)SC#N
219.218
304.34
97%
White
885266-67-1
MFCD04973974
FDQWHYSXVRDVMV-UHFFFAOYSA-N
(4-nitro-1H-indol-3-yl) thiocyanate
24720942
Solid

Chemical Identifiers

885266-67-1
219.218
FDQWHYSXVRDVMV-UHFFFAOYSA-N
24720942
C1=CC2=C(C(=C1)[N+](=O)[O-])C(=CN2)SC#N
C9H5N3O2S
MFCD04973974
4-nitro-1h-indol-3-yl sulfanyl formonitrile
(4-nitro-1H-indol-3-yl) thiocyanate
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents