Please login to your online account to display your discounted pricing

1,2-Bis(trimethylsilyloxy)ethane, 98%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  209580100

 View more versions of this product

Catalog No. AC209580100

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥97.5%
Formula Weight 206.43
Quantity 10g
Color Clear colorless to faint yellow
CAS Max % 100.0
Refractive Index 1.4020 to 1.4040 (20°C, 589nm)
Chemical Name or Material 1, 2-Bis(trimethylsilyloxy)ethane
Packaging Glass bottle
Molecular Formula C8H22O2Si2
MDL Number MFCD00009640
CAS 7381-30-8
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula (CH3)3SiOCH2CH2OSi(CH3)3
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1, 2-Bis(trimethylsilyloxy)ethane
CAS 7381-30-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H22O2Si2
Linear Formula (CH3)3SiOCH2CH2OSi(CH3)3
Formula Weight 206.43
MDL Number MFCD00009640
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.
Flammable liquid and vapor.

SDS & Safety Documents