missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Bis(trimethylsiloxy)ethane, 98%

$48.89 - $769.51

Chemical Identifiers

CAS 7381-30-8
Molecular Formula C8H22O2Si2
Molecular Weight (g/mol) 206.432
MDL Number MFCD00009640
InChI Key JGWFUSVYECJQDT-UHFFFAOYSA-N
Synonym 1,2-bis trimethylsiloxy ethane, 1,2-bis trimethylsilyloxy ethane, 2,2,7,7-tetramethyl-3,6-dioxa-2,7-disilaoctane, ethylene glycol bis trimethylsilyl ether, ethylenedioxybis trimethylsilane, 3,6-dioxa-2,7-disilaoctane, 2,2,7,7-tetramethyl, 1,2-bis-trimethylsilyloxy ethane, 1,2-bis-trimethylsilanyloxy-ethane, ethylene glycol, di-tms, acmc-1bh50
PubChem CID 81858
IUPAC Name trimethyl(2-trimethylsilyloxyethoxy)silane
SMILES C[Si](C)(C)OCCO[Si](C)(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6385709 View Documents Alfa Aesar™
H6385709
10g Each for $48.89
 
AAH6385718 View Documents Alfa Aesar™
H6385718
50g Each for $175.80
 
AAH6385730 View Documents Alfa Aesar™
H6385730
250g Each for $769.51
 
Specifications

Chemical Identifiers

7381-30-8
206.432
JGWFUSVYECJQDT-UHFFFAOYSA-N
81858
C[Si](C)(C)OCCO[Si](C)(C)C
C8H22O2Si2
MFCD00009640
1,2-bis trimethylsiloxy ethane, 1,2-bis trimethylsilyloxy ethane, 2,2,7,7-tetramethyl-3,6-dioxa-2,7-disilaoctane, ethylene glycol bis trimethylsilyl ether, ethylenedioxybis trimethylsilane, 3,6-dioxa-2,7-disilaoctane, 2,2,7,7-tetramethyl, 1,2-bis-trimethylsilyloxy ethane, 1,2-bis-trimethylsilanyloxy-ethane, ethylene glycol, di-tms, acmc-1bh50
trimethyl(2-trimethylsilyloxyethoxy)silane

Specifications

7381-30-8
(CH3)3SiOCH2CH2OSi(CH3)3
UN1993
1,2-bis trimethylsiloxy ethane, 1,2-bis trimethylsilyloxy ethane, 2,2,7,7-tetramethyl-3,6-dioxa-2,7-disilaoctane, ethylene glycol bis trimethylsilyl ether, ethylenedioxybis trimethylsilane, 3,6-dioxa-2,7-disilaoctane, 2,2,7,7-tetramethyl, 1,2-bis-trimethylsilyloxy ethane, 1,2-bis-trimethylsilanyloxy-ethane, ethylene glycol, di-tms, acmc-1bh50
C[Si](C)(C)OCCO[Si](C)(C)C
206.432
206.43
165°C to 171°C
51°C (124°F)
C8H22O2Si2
MFCD00009640
1744217
JGWFUSVYECJQDT-UHFFFAOYSA-N
trimethyl(2-trimethylsilyloxyethoxy)silane
81858
98%
10g
1,2-Bis(trimethylsiloxy)ethane
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.