missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Bis(triethoxysilyl)ethane, 95%

$21.91 - $92.38

Chemical Identifiers

CAS 16068-37-4
Molecular Formula C14H34O6Si2
Molecular Weight (g/mol) 354.59
MDL Number MFCD00239362
InChI Key IZRJPHXTEXTLHY-UHFFFAOYSA-N
Synonym 1,2-bis triethoxysilyl ethane, 4,4,7,7-tetraethoxy-3,8-dioxa-4,7-disiladecane, 3,8-dioxa-4,7-disiladecane, 4,4,7,7-tetraethoxy, ethylenebis triethoxysilane, bis triethoxysilyl ethane, hexaethoxydisilethylene, bis 1,2-triethoxysilyl ethane, 1,2-bis triethoxysilyl ethane,, triethoxysilyl ethyl triethoxysilane, triethoxy 2-triethoxysilylethyl silane
PubChem CID 85266
IUPAC Name triethoxy(2-triethoxysilylethyl)silane
SMILES CCO[Si](CC[Si](OCC)(OCC)OCC)(OCC)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60530AA View Documents Alfa Aesar™
H60530AA
5mL Each for $21.91
 
AAH60530AC View Documents Alfa Aesar™
H60530AC
25mL Each for $92.38
 
Description

Description

Applications
1,2-Bis(triethoxysilyl)ethane is used as coating auxiliary agents, leather auxiliary agents, plastic auxiliary agents, surfactants and textile auxiliary agents. It is used to prepare mesoporous organosilica materials.

Solubility
Soluble in alcohol, aromatic and aliphatic hydrocarbons. Hydrolyzes with water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
Specifications

Chemical Identifiers

16068-37-4
354.59
IZRJPHXTEXTLHY-UHFFFAOYSA-N
85266
CCO[Si](CC[Si](OCC)(OCC)OCC)(OCC)OCC
C14H34O6Si2
MFCD00239362
1,2-bis triethoxysilyl ethane, 4,4,7,7-tetraethoxy-3,8-dioxa-4,7-disiladecane, 3,8-dioxa-4,7-disiladecane, 4,4,7,7-tetraethoxy, ethylenebis triethoxysilane, bis triethoxysilyl ethane, hexaethoxydisilethylene, bis 1,2-triethoxysilyl ethane, 1,2-bis triethoxysilyl ethane,, triethoxysilyl ethyl triethoxysilane, triethoxy 2-triethoxysilylethyl silane
triethoxy(2-triethoxysilylethyl)silane

Specifications

16068-37-4
[-CH2Si(OC2H5)3]2
1,2-bis triethoxysilyl ethane, 4,4,7,7-tetraethoxy-3,8-dioxa-4,7-disiladecane, 3,8-dioxa-4,7-disiladecane, 4,4,7,7-tetraethoxy, ethylenebis triethoxysilane, bis triethoxysilyl ethane, hexaethoxydisilethylene, bis 1,2-triethoxysilyl ethane, 1,2-bis triethoxysilyl ethane,, triethoxysilyl ethyl triethoxysilane, triethoxy 2-triethoxysilylethyl silane
CCO[Si](CC[Si](OCC)(OCC)OCC)(OCC)OCC
354.59
354.59
119°C
113°C (235°F)
C14H34O6Si2
MFCD00239362
IZRJPHXTEXTLHY-UHFFFAOYSA-N
triethoxy(2-triethoxysilylethyl)silane
85266
95%
5mL
1,2-Bis(triethoxysilyl)ethane
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : JG7755000

TSCA : TSCA