missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2-Bis(trichlorosilyl)ethane 97.0+%, TCI America™
$61.38 - $296.97
Chemical Identifiers
CAS | 2504-64-5 |
---|---|
Molecular Formula | C2H4Cl6Si2 |
Molecular Weight (g/mol) | 296.924 |
MDL Number | MFCD00013601 |
InChI Key | WDVUXWDZTPZIIE-UHFFFAOYSA-N |
PubChem CID | 75631 |
IUPAC Name | trichloro(2-trichlorosilylethyl)silane |
SMILES | C(C[Si](Cl)(Cl)Cl)[Si](Cl)(Cl)Cl |
Chemical Identifiers
2504-64-5 | |
296.924 | |
WDVUXWDZTPZIIE-UHFFFAOYSA-N | |
trichloro(2-trichlorosilylethyl)silane |
C2H4Cl6Si2 | |
MFCD00013601 | |
75631 | |
C(C[Si](Cl)(Cl)Cl)[Si](Cl)(Cl)Cl |
Specifications
2504-64-5 | |
C2H4Cl6Si2 | |
5 g | |
WDVUXWDZTPZIIE-UHFFFAOYSA-N | |
trichloro(2-trichlorosilylethyl)silane | |
75631 | |
≥97.0% (T) | |
1,2-Bis(trichlorosilyl)ethane |
White | |
MFCD00013601 | |
2987 | |
C(C[Si](Cl)(Cl)Cl)[Si](Cl)(Cl)Cl | |
296.924 | |
296.92 | |
Crystalline Lumps |
Safety and Handling
TSCA : Yes