Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Bis(diphenylphosphino)benzene, 98%

$145.46 - $145.46

Chemical Identifiers

Chemical Name or Material 1,2-Bis(diphenylphosphino)benzene
CAS 13991-08-7
Molecular Formula C26H22P2
Formula Weight 446.47
MDL Number MFCD00014081
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2110603

  SDS
alfa aesar™
B2110603
1g Each for $145.46
Description & Specifications

Specifications

Formula Weight 446.47
Chemical Name or Material 1,2-Bis(diphenylphosphino)benzene
Assay 98%
Molecular Formula C26H22P2
MDL Number MFCD00014081
CAS 13991-08-7
Melting Point 185° to 189°C
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Applications
It acts as a ligand for Baeyer-Villiger Oxidation, cycloadditions and reductions.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.