Please login to your online account to display your discounted pricing

1,2-Bis(dipentafluorophenylphosphino)ethane, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  363700010

 View more versions of this product

Catalog No. AC363700010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 1, 2-Bis(dipentafluorophenylphosphino)ethane
CAS 76858-94-1
Infrared Spectrum Authentic
Physical Form Crystalline Powder
Assay ≥94% (HPLC)
Assay Percent Range 95%
Molecular Formula C26H4F20P2
Formula Weight 758.26
Color White to light beige
Melting Point 190° to 195°C
Quantity 1g

Specifications

Chemical Name or Material 1, 2-Bis(dipentafluorophenylphosphino)ethane
Assay ≥94% (HPLC)
Assay Percent Range 95%
Molecular Formula C26H4F20P2
Formula Weight 758.26