Please login to your online account to display your discounted pricing

1,2-Bis(dipentafluorophenylphosphino)ethane, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 2-Bis(dipentafluorophenylphosphino)ethane
Assay ≥94% (HPLC)
Assay Percent Range 95%
Molecular Formula C26H4F20P2
Formula Weight 758.26
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC363700010

  SDS
thermo fisher scientific
363700010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC363700050

  SDS
thermo fisher scientific
AC363700050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94% (HPLC)
Physical Form Crystalline Powder
Formula Weight 758.26
Quantity 1g
Color White to light beige
Melting Point 190° to 195°C
Molecular Formula C26H4F20P2
CAS 76858-94-1
Infrared Spectrum Authentic
Chemical Name or Material 1, 2-Bis(dipentafluorophenylphosphino)ethane
Assay Percent Range 95%