Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Bis(dipentafluorophenylphosphino)ethane, 99%

Chemical Identifiers

Chemical Name or Material 1,2-Bis(dipentafluorophenylphosphino)ethane
CAS 76858-94-1
Linear Formula (C6F5)2PCH2CH2P(C6F5)2
Formula Weight 758.23
MDL Number MFCD00058841
UN Number UN3464
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3927603

  SDS
alfa aesar™
3927603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3927606

  SDS
alfa aesar™
3927606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 758.23
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1,2-Bis(dipentafluorophenylphosphino)ethane
Physical Form Crystalline
Assay 99%
Linear Formula (C6F5)2PCH2CH2P(C6F5)2
MDL Number MFCD00058841
CAS 76858-94-1
UN Number UN3464
Color White
Melting Point 185° to 190°C
Health Hazard 1 Danger
Quantity 1g
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.