missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Bis(dimethylsilyl)benzene, 98%

Manufacturer:  Alfa Aesar™ 4375502

 View more versions of this product

Catalog No. AA4375502



Specifications

Specifications

1,2-Bis (dimethylsilyl)benzene
70°C (158°F)
101°C (13mmHg)
17985-72-7
MFCD00142462
MUUXBTFQEXVEEI-UHFFFAOYSA-N
[2-(dimethyl-$l^{3}-silanyl)phenyl]-dimethylsilicon
6364986
Liquid
0.898g/mL
1.5140
0.5g
C10H16Si2
1,2-bis dimethylsilyl benzene
C[Si](C)C1=CC=CC=C1[Si](C)C
192.408
194.43
98%

Chemical Identifiers

17985-72-7
192.408
MUUXBTFQEXVEEI-UHFFFAOYSA-N
6364986
C[Si](C)C1=CC=CC=C1[Si](C)C
C10H16Si2
MFCD00142462
1,2-bis dimethylsilyl benzene
[2-(dimethyl-$l^{3}-silanyl)phenyl]-dimethylsilicon
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents