Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Bis(dimethylphosphino)ethane, 97%

Chemical Identifiers

Chemical Name or Material 1,2-Bis(dimethylphosphino)ethane
CAS 23936-60-9
Molecular Formula C6H16P2
Linear Formula (CH3)2PCH2CH2P(CH3)2
Formula Weight 150.14
MDL Number MFCD00008511
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2105101

  SDS
alfa aesar™
B2105101
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2105103

  SDS
alfa aesar™
B2105103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 150.14
Flash Point −17°C (1.4°F)
Chemical Name or Material 1,2-Bis(dimethylphosphino)ethane
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 23936-60-9
Linear Formula (CH3)2PCH2CH2P(CH3)2
Health Hazard 3 GHS P Statement
P210-P261-P280g-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 97%
UN Number UN1993
Molecular Formula C6H16P2
Quantity 0.25g
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Boiling Point 180°C
Odor Recognizable
MDL Number MFCD00008511
Melting Point 179° to 180°C
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P210-P261-P280g-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.