Learn More
1,2-Bis(diethylphosphino)ethane, 98%, Thermo Scientific Chemicals
$169.29 - $649.30
Chemical Identifiers
CAS | 6411-21-8 |
---|---|
Molecular Formula | C10H24P2 |
Molecular Weight (g/mol) | 206.25 |
MDL Number | MFCD00015175 |
InChI Key | MIOCUERTSIJEDP-UHFFFAOYSA-N |
Synonym | 1,2-bis diethylphosphino ethane, bis 1,2-diethylphosphino ethane, 1,2-bis-diethylphosphino-ethane, acmc-1bbse, ethylenebis diethylphosphine, chembl70363, 1,2 bis diethylphosphino ethane, 1,2-bis-diethylphosphino ethane |
PubChem CID | 138745 |
SMILES | CCP(CC)CCP(CC)CC |
Description
It is used in chemical reaearch, organic Intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Chemical Identifiers
6411-21-8 | |
206.25 | |
MIOCUERTSIJEDP-UHFFFAOYSA-N | |
138745 |
C10H24P2 | |
MFCD00015175 | |
1,2-bis diethylphosphino ethane, bis 1,2-diethylphosphino ethane, 1,2-bis-diethylphosphino-ethane, acmc-1bbse, ethylenebis diethylphosphine, chembl70363, 1,2 bis diethylphosphino ethane, 1,2-bis-diethylphosphino ethane | |
CCP(CC)CCP(CC)CC |
Specifications
6411-21-8 | |
0.884 g/mL | |
Pungent | |
(CH3CH2)2PCH2CH2P(CH2CH3)2 | |
1 g | |
Air Sensitive | |
It is soluble in organic solvents. | |
CCP(CC)CCP(CC)CC | |
206.25 | |
206.25 | |
Liquid |
Colorless | |
124°C to 126°C (10 mmHg) | |
C10H24P2 | |
MFCD00015175 | |
UN2845 | |
1,2-bis diethylphosphino ethane, bis 1,2-diethylphosphino ethane, 1,2-bis-diethylphosphino-ethane, acmc-1bbse, ethylenebis diethylphosphine, chembl70363, 1,2 bis diethylphosphino ethane, 1,2-bis-diethylphosphino ethane | |
MIOCUERTSIJEDP-UHFFFAOYSA-N | |
2-diethylphosphanylethyl(diethyl)phosphane | |
138745 | |
98% | |
1,2-Bis(diethylphosphino)ethane |
Safety and Handling
GHS H Statement
H250-H252-H301-H311-H331
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
P210-P222-P261-P264b-P270-P271-P280-P301+P312-P302+P334-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H250-H302+H312-H315-H319-H335
DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
TSCA : No
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only