Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Bis(dicyclohexylphosphino)ethane, 98%

Chemical Identifiers

Chemical Name or Material 1,2-Bis(dicyclohexylphosphino)ethane
CAS 23743-26-2
Formula Weight 422.61
MDL Number MFCD00015521
UN Number UN1325
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3926902

  SDS
alfa aesar™
3926902
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3926904

  SDS
alfa aesar™
3926904
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 422.61
Chemical Name or Material 1,2-Bis(dicyclohexylphosphino)ethane
Physical Form Crystalline
Assay 98%
MDL Number MFCD00015521
CAS 23743-26-2
UN Number UN1325
Health Hazard 1 Danger
Quantity 0.5g
Sensitivity Air Sensitive
Health Hazard 2 GHS H Statement
H228
Flammable solid.
Health Hazard 3 GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
Storage Note 1
  • Packaged under argon
DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

  • Packaged under argon

Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

GHS H Statement
H228
Flammable solid.

  • Packaged under argon