Learn More
1,2-Bis(chlorodimethylsilyl)ethane, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 313260250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 2-Bis(chlorodimethylsilyl)ethane | |
13528-93-3 | |
100.0 | |
198.0°C to 200.0°C | |
Authentic | |
Glass bottle | |
[-CH2Si(CH3)2Cl]2 | |
25 g | |
1,2-bis chlorodimethylsilyl ethane, ethylenebis chlorodimethylsilane, silane, 1,2-ethanediylbis chlorodimethyl, unii-b4ocj0e6j1, 1,1,4,4-tetramethyl-1,4-dichlorodisilethylene, chloro 2-chlorodimethylsilyl ethyl dimethylsilane, b4ocj0e6j1, 2,5-dichloro-2,5-dimethyl-2,5-disilahexane, 1,1,4,4-tetramethyl-1,4-dichloro-disylethylene | |
VGQOKOYKFDUPPJ-UHFFFAOYSA-N | |
chloro-[2-[chloro(dimethyl)silyl]ethyl]-dimethylsilane | |
83552 | |
96% |
96% | |
95.0 | |
35.0°C to 38.0°C | |
65°C | |
96% | |
C6H16Cl2Si2 | |
MFCD00000505 | |
15, 1251 | |
Solubility in water: reacts. | |
C[Si](C)(Cl)CC[Si](C)(C)Cl | |
215.26 | |
215.27 | |
Low Melting Solid |
Chemical Identifiers
13528-93-3 | |
215.26 | |
VGQOKOYKFDUPPJ-UHFFFAOYSA-N | |
83552 | |
C[Si](C)(Cl)CC[Si](C)(C)Cl |
C6H16Cl2Si2 | |
MFCD00000505 | |
1,2-bis chlorodimethylsilyl ethane, ethylenebis chlorodimethylsilane, silane, 1,2-ethanediylbis chlorodimethyl, unii-b4ocj0e6j1, 1,1,4,4-tetramethyl-1,4-dichlorodisilethylene, chloro 2-chlorodimethylsilyl ethyl dimethylsilane, b4ocj0e6j1, 2,5-dichloro-2,5-dimethyl-2,5-disilahexane, 1,1,4,4-tetramethyl-1,4-dichloro-disylethylene | |
chloro-[2-[chloro(dimethyl)silyl]ethyl]-dimethylsilane |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 236-871-0
TSCA : TSCA
RUO – Research Use Only