Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Bis(carboxymethylthio)ethane, 97%

$76.82 - $1,037.47

Chemical Identifiers

Chemical Name or Material 1,2-Bis(carboxymethylthio)ethane
CAS 7244-02-2
Molecular Formula (CH2SCH2CO2H)2
Formula Weight 210.26
MDL Number MFCD00021771
Synonym 1,2-Bis(carboxymethylmercapto)ethane; 1,2-Ethanediylbis(thioacetic acid)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2107406

 
alfa aesar™
B2107406
5g Each for $76.82

AAB2107422

 
alfa aesar™
B2107422
100g Each for $1,037.47
Description & Specifications

Specifications

Formula Weight 210.26
Chemical Name or Material 1,2-Bis(carboxymethylthio)ethane
Assay 97%
Molecular Formula (CH2SCH2CO2H)2
MDL Number MFCD00021771
CAS 7244-02-2
Melting Point 110°C
Synonym 1,2-Bis(carboxymethylmercapto)ethane; 1,2-Ethanediylbis(thioacetic acid)
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.