missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Bis[(2S,5S)-2,5-diphenyl-1-phospholanyl]ethane, 97+%, Alfa Aesar™

$271.42 - $936.07

Chemical Identifiers

CAS 824395-67-7
Molecular Formula C34H36P2
Molecular Weight (g/mol) 506.61
MDL Number MFCD07369028
InChI Key VHHAZLMVLLIMHT-CUPIEXAXSA-N
Synonym +-1,2-bis 2s,5s-2,5-diphenylphospholano ethane, 1,2-bis 2s,5s-2,5-diphenylphospholan-1-yl ethane, 1,1'-ethylenebis 2alpha,5beta-diphenyltetrahydro-1h-phosphole, +-1,2-bis 2s,5s-2,5-diphenylphospholano ethane, kanata purity, 2s,5s-1-2-2s,5s-2,5-diphenylphospholan-1-yl ethyl-2,5-diphenylphospholane
PubChem CID 16218207
IUPAC Name (2S,5S)-1-[2-[(2S,5S)-2,5-diphenylphospholan-1-yl]ethyl]-2,5-diphenylphospholane
SMILES C1CC(P(C1C2=CC=CC=C2)CCP3C(CCC3C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60766MD View Documents Alfa Aesar
H60766MD
250mg Each for $271.42
 
AAH6076603 View Documents Alfa Aesar
H6076603
1g Each for $936.07
 
Description

Description

Applications
It is a DuPhos and BPE ligands which are highly efficient privileged ligands. It is a chiral phosphine ligand used in the asymmetric hydrogenation of allylic sulfoxides.

Solubility
Sparingly Soluble (3.2E-5 g/L) (25°C).

Notes
Incompatible with strong oxidizing agents. Store under room temperature. Air sensitive.
Specifications

Chemical Identifiers

824395-67-7
506.61
VHHAZLMVLLIMHT-CUPIEXAXSA-N
16218207
C1CC(P(C1C2=CC=CC=C2)CCP3C(CCC3C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6
C34H36P2
MFCD07369028
+-1,2-bis 2s,5s-2,5-diphenylphospholano ethane, 1,2-bis 2s,5s-2,5-diphenylphospholan-1-yl ethane, 1,1'-ethylenebis 2alpha,5beta-diphenyltetrahydro-1h-phosphole, +-1,2-bis 2s,5s-2,5-diphenylphospholano ethane, kanata purity, 2s,5s-1-2-2s,5s-2,5-diphenylphospholan-1-yl ethyl-2,5-diphenylphospholane
(2S,5S)-1-[2-[(2S,5S)-2,5-diphenylphospholan-1-yl]ethyl]-2,5-diphenylphospholane

Specifications

143°C to 147°C
824395-67-7
MFCD07369028
VHHAZLMVLLIMHT-CUPIEXAXSA-N
(2S,5S)-1-[2-[(2S,5S)-2,5-diphenylphospholan-1-yl]ethyl]-2,5-diphenylphospholane
16218207
≥97%
250mg
C34H36P2
+-1,2-bis 2s,5s-2,5-diphenylphospholano ethane, 1,2-bis 2s,5s-2,5-diphenylphospholan-1-yl ethane, 1,1'-ethylenebis 2alpha,5beta-diphenyltetrahydro-1h-phosphole, +-1,2-bis 2s,5s-2,5-diphenylphospholano ethane, kanata purity, 2s,5s-1-2-2s,5s-2,5-diphenylphospholan-1-yl ethyl-2,5-diphenylphospholane
C1CC(P(C1C2=CC=CC=C2)CCP3C(CCC3C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6
506.61
506.6
1,2-Bis[(2S,5S)-2,5-diphenyl-1-phospholanyl]ethane