missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC435231000

 View more versions of this product

Catalog No. AC435231000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

1,2-Bis((2S,5S)-2,5-Dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate
(1H-NMR) Consistent with structure, (31P-NMR) Consistent with structure, NMR (19F-NMR), Consistent with structure
152.0°C to 156.0°C
213343-65-8
100.0
BF4·HRh
LKVIVYCYPYTYSO-ASDDUFFSSA-N
(1Z,5Z)-cycloocta-1,5-diene;(2S,5S)-1-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]ethyl]-2,5-dimethylphospholane;rhodium;tetrafluoroborate
12964417
Crystalline Powder
Authentic
Brown to Orange
100mg
96.0
97%
--1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene rhodium i tetrafluoroborate, --1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene-rhodium i tetrafluoroborate, 1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene rhodium i tetrafluoroborate, s,s-me-bpe-rh
[B-](F)(F)(F)F.CC1CCC(P1CCP2C(CCC2C)C)C.C1CC=CCCC=C1.[Rh]
556.21
556.21
97%

Chemical Identifiers

213343-65-8
556.21
--1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene rhodium i tetrafluoroborate, --1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene-rhodium i tetrafluoroborate, 1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene rhodium i tetrafluoroborate, s,s-me-bpe-rh
(1Z,5Z)-cycloocta-1,5-diene;(2S,5S)-1-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]ethyl]-2,5-dimethylphospholane;rhodium;tetrafluoroborate
BF4·HRh
LKVIVYCYPYTYSO-ASDDUFFSSA-N
12964417
[B-](F)(F)(F)F.CC1CCC(P1CCP2C(CCC2C)C)C.C1CC=CCCC=C1.[Rh]
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents