missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]ethane, 97+%, Alfa Aesar™

$143.88 - $498.11

Chemical Identifiers

CAS 136779-26-5
Molecular Formula C14H28P2
Molecular Weight (g/mol) 258.326
MDL Number MFCD01073771
InChI Key IRCDUOCGSIGEAI-XUXIUFHCSA-N
Synonym s,s-me-bpe, me-bpe, s,s, unii-23b6d1oycy, 23b6d1oycy, --1,2-bis 2s,5s-2,5-dimethylphospholano ethane, --1,2-bis 2s,5s-2,5-dimethylphospholano ethane, s,s-me-bpe mi, s,s-me-bpe,-, 2s,5s-1-2-2s,5s-2,5-dimethylphospholan-1-yl ethyl-2,5-dimethylphospholane, 2s,5s-1-2-2s,5s-2,5-dimethylphospholan-1-yl ethyl-2,5-dimethylphospholane
PubChem CID 5702644
IUPAC Name (2S,5S)-1-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]ethyl]-2,5-dimethylphospholane
SMILES CC1CCC(P1CCP2C(CCC2C)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60265MD View Documents Alfa Aesar
H60265MD
250mg Each for $143.88
 
AAH6026503 View Documents Alfa Aesar
H6026503
1g Each for $498.11
 
Specifications

Chemical Identifiers

136779-26-5
258.326
IRCDUOCGSIGEAI-XUXIUFHCSA-N
5702644
CC1CCC(P1CCP2C(CCC2C)C)C
C14H28P2
MFCD01073771
s,s-me-bpe, me-bpe, s,s, unii-23b6d1oycy, 23b6d1oycy, --1,2-bis 2s,5s-2,5-dimethylphospholano ethane, --1,2-bis 2s,5s-2,5-dimethylphospholano ethane, s,s-me-bpe mi, s,s-me-bpe,-, 2s,5s-1-2-2s,5s-2,5-dimethylphospholan-1-yl ethyl-2,5-dimethylphospholane, 2s,5s-1-2-2s,5s-2,5-dimethylphospholan-1-yl ethyl-2,5-dimethylphospholane
(2S,5S)-1-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]ethyl]-2,5-dimethylphospholane

Specifications

64°C to 67°C (0.06mmHg)
136779-26-5
MFCD01073771
s,s-me-bpe, me-bpe, s,s, unii-23b6d1oycy, 23b6d1oycy, --1,2-bis 2s,5s-2,5-dimethylphospholano ethane, --1,2-bis 2s,5s-2,5-dimethylphospholano ethane, s,s-me-bpe mi, s,s-me-bpe,-, 2s,5s-1-2-2s,5s-2,5-dimethylphospholan-1-yl ethyl-2,5-dimethylphospholane, 2s,5s-1-2-2s,5s-2,5-dimethylphospholan-1-yl ethyl-2,5-dimethylphospholane
CC1CCC(P1CCP2C(CCC2C)C)C
258.326
258.32
1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]ethane
250mg
C14H28P2
UN2845
IRCDUOCGSIGEAI-XUXIUFHCSA-N
(2S,5S)-1-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]ethyl]-2,5-dimethylphospholane
5702644
≥97%
Safety and Handling

Safety and Handling

GHS H Statement
H250-H252-H315-H319-H335
Catches fire spontaneously if exposed to air.
Self-heating in large quantities; may catch fire.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P222-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not allow contact with air.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.