missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene, 97+%, Thermo Scientific Chemicals
$211.44 - $806.15
Chemical Identifiers
CAS | 136735-95-0 |
---|---|
Molecular Formula | C18H28P2 |
Molecular Weight (g/mol) | 306.37 |
MDL Number | MFCD00142322 |
InChI Key | AJNZWRKTWQLAJK-VGWMRTNUSA-N |
Synonym | s,s-me-duphos, methyl-duphos, s,s, s,s-methyl-duphos, unii-hl745j99sb, +-1,2-bis 2s,5s-2,5-dimethylphospholano benzene, phospholane, 1,1'-1,2-phenylene bis 2,5-dimethyl-, 2s,2's,5s,5's, s,s-me-duphos mi, s,s-methyl-duphos, +, 2s,5s-1-2-2s,5s-2,5-dimethylphospholan-1-yl phenyl-2,5-dimethylphospholane |
PubChem CID | 2734553 |
IUPAC Name | (2S,5S)-1-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethylphospholane |
SMILES | CC1CCC(P1C2=CC=CC=C2P3C(CCC3C)C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH60647MD
|
Thermo Scientific Chemicals
H60647MD |
250 mg |
N/A
|
|
|||||
AAH6064703
|
Thermo Scientific Chemicals
H6064703 |
1 g |
N/A
|
|
|||||
Description
It is a DuPhos and BPE ligands which are highly efficient privileged ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a DuPhos and BPE ligands which are highly efficient privileged ligands.
Solubility
Insoluble in water.
Notes
Air sensitive. Store at room temperature.
Chemical Identifiers
136735-95-0 | |
306.37 | |
AJNZWRKTWQLAJK-VGWMRTNUSA-N | |
2734553 | |
CC1CCC(P1C2=CC=CC=C2P3C(CCC3C)C)C |
C18H28P2 | |
MFCD00142322 | |
s,s-me-duphos, methyl-duphos, s,s, s,s-methyl-duphos, unii-hl745j99sb, +-1,2-bis 2s,5s-2,5-dimethylphospholano benzene, phospholane, 1,1'-1,2-phenylene bis 2,5-dimethyl-, 2s,2's,5s,5's, s,s-me-duphos mi, s,s-methyl-duphos, +, 2s,5s-1-2-2s,5s-2,5-dimethylphospholan-1-yl phenyl-2,5-dimethylphospholane | |
(2S,5S)-1-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethylphospholane |
Specifications
136735-95-0 | |
C18H28P2 | |
250 mg | |
Air Sensitive | |
Insoluble in water. | |
CC1CCC(P1C2=CC=CC=C2P3C(CCC3C)C)C | |
306.37 | |
306.36 | |
1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene |
66°C to 76°C | |
MFCD00142322 | |
4810602 | |
s,s-me-duphos, methyl-duphos, s,s, s,s-methyl-duphos, unii-hl745j99sb, +-1,2-bis 2s,5s-2,5-dimethylphospholano benzene, phospholane, 1,1'-1,2-phenylene bis 2,5-dimethyl-, 2s,2's,5s,5's, s,s-me-duphos mi, s,s-methyl-duphos, +, 2s,5s-1-2-2s,5s-2,5-dimethylphospholan-1-yl phenyl-2,5-dimethylphospholane | |
AJNZWRKTWQLAJK-VGWMRTNUSA-N | |
(2S,5S)-1-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethylphospholane | |
2734553 | |
≥97% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only