Please login to your online account to display your discounted pricing

1,2-Bis((2S,5S)-2,5-diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1,2-Bis((2S,5S)-2,5-Diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate
CAS 1000393-36-1
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C34H56P2Rh·BF4
Formula Weight 716.48
MDL Number MFCD07369040
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC435210500

  SDS
thermo fisher scientific
AC435210500
50mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC435212500

  SDS
thermo fisher scientific
AC435212500
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 716.48
Quantity 50mg
Color Yellow to orange to red to brown
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 1,2-Bis((2S,5S)-2,5-Diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C34H56P2Rh·BF4
MDL Number MFCD07369040
CAS 1000393-36-1
Elemental Analysis 54.7 to 59.4% C, 7.6 to 8.2% H
NMR (1H-NMR) Consistent with structure, (31P-NMR) Consistent with structure, NMR (19F-NMR), Consistent with structure