missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]benzene, 97+%, Alfa Aesar™

$211.27 - $697.04

Chemical Identifiers

CAS 147253-69-8
Molecular Formula C26H44P2
Molecular Weight (g/mol) 418.586
MDL Number MFCD02684550
InChI Key RBVGOQHQBUPSGX-ZJZGAYNASA-N
Synonym s,s-i-pr-duphos, rel-1,2-bis 2s,5s-2,5-diisopropylphospholan-1-yl benzene, s,s-ipr-duphos, 1,2-bis 2s,5s-2,5-diisopropylphospholan-1-yl benzene, --1,2-bis 2s,5s-2,5-di-iso-propylphospholano benzene, --1,2-bis 2s,5s-2,5-di-i-propylphospholano benzene, --1,2-bis 2s,5s-2,5-diisopropylphospholano benzene, kanata purity, 2s,5s-1-2-2s,5s-2,5-diisopropylphospholan-1-yl phenyl-2,5-diisopropylphospholane
PubChem CID 11520495
IUPAC Name (2S,5S)-1-[2-[(2S,5S)-2,5-di(propan-2-yl)phospholan-1-yl]phenyl]-2,5-di(propan-2-yl)phospholane
SMILES CC(C)C1CCC(P1C2=CC=CC=C2P3C(CCC3C(C)C)C(C)C)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60407MD View Documents Alfa Aesar
H60407MD
250mg Each for $211.27
 
AAH6040703 View Documents Alfa Aesar
H6040703
1g Each for $697.04
 
Specifications

Chemical Identifiers

147253-69-8
418.586
RBVGOQHQBUPSGX-ZJZGAYNASA-N
11520495
CC(C)C1CCC(P1C2=CC=CC=C2P3C(CCC3C(C)C)C(C)C)C(C)C
C26H44P2
MFCD02684550
s,s-i-pr-duphos, rel-1,2-bis 2s,5s-2,5-diisopropylphospholan-1-yl benzene, s,s-ipr-duphos, 1,2-bis 2s,5s-2,5-diisopropylphospholan-1-yl benzene, --1,2-bis 2s,5s-2,5-di-iso-propylphospholano benzene, --1,2-bis 2s,5s-2,5-di-i-propylphospholano benzene, --1,2-bis 2s,5s-2,5-diisopropylphospholano benzene, kanata purity, 2s,5s-1-2-2s,5s-2,5-diisopropylphospholan-1-yl phenyl-2,5-diisopropylphospholane
(2S,5S)-1-[2-[(2S,5S)-2,5-di(propan-2-yl)phospholan-1-yl]phenyl]-2,5-di(propan-2-yl)phospholane

Specifications

−103° (c=1 in Chloroform)
250mg
C26H44P2
s,s-i-pr-duphos, rel-1,2-bis 2s,5s-2,5-diisopropylphospholan-1-yl benzene, s,s-ipr-duphos, 1,2-bis 2s,5s-2,5-diisopropylphospholan-1-yl benzene, --1,2-bis 2s,5s-2,5-di-iso-propylphospholano benzene, --1,2-bis 2s,5s-2,5-di-i-propylphospholano benzene, --1,2-bis 2s,5s-2,5-diisopropylphospholano benzene, kanata purity, 2s,5s-1-2-2s,5s-2,5-diisopropylphospholan-1-yl phenyl-2,5-diisopropylphospholane
CC(C)C1CCC(P1C2=CC=CC=C2P3C(CCC3C(C)C)C(C)C)C(C)C
418.586
418.58
1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]benzene
26°C to 43°C
147253-69-8
MFCD02684550
RBVGOQHQBUPSGX-ZJZGAYNASA-N
(2S,5S)-1-[2-[(2S,5S)-2,5-di(propan-2-yl)phospholan-1-yl]phenyl]-2,5-di(propan-2-yl)phospholane
11520495
≥97%