missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals 1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 435192500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,2-Bis((2S,5S)-2,5-Diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate | |
96.0 | |
Brown to Orange | |
97% | |
C30H48P2Rh·BF4 | |
(1H-NMR) Consistent with structure, (31P-NMR) Consistent with structure, NMR (19F-NMR), Consistent with structure | |
250mg | |
XIOPHSBHVOCZMM-ZCTOJWETSA-N | |
(1Z,5Z)-cycloocta-1,5-diene;(2S,5S)-1-[2-[(2S,5S)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane;rhodium;tetrafluoroborate | |
11006841 | |
Powder |
213343-64-7 | |
100.0 | |
52.4 to 56.8% C | |
Authentic | |
MFCD01862465 | |
Glass bottle | |
s,s-et-duphos-rh, +-1,2-bis 2s,5s-2,5-diethylphospholano benzene cyclooctadiene rhodium i tetrafluoroborate, +-1,2-bis 2s,5s-2,5-diethylphospholano benzene cyclooctadiene rhodium l tetrafluoroborate | |
[B-](F)(F)(F)F.CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC.C1CC=CCCC=C1.[Rh] | |
660.37 | |
660.37 | |
97% |
Chemical Identifiers
213343-64-7 | |
660.37 | |
XIOPHSBHVOCZMM-ZCTOJWETSA-N | |
11006841 | |
[B-](F)(F)(F)F.CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC.C1CC=CCCC=C1.[Rh] |
C30H48P2Rh·BF4 | |
MFCD01862465 | |
s,s-et-duphos-rh, +-1,2-bis 2s,5s-2,5-diethylphospholano benzene cyclooctadiene rhodium i tetrafluoroborate, +-1,2-bis 2s,5s-2,5-diethylphospholano benzene cyclooctadiene rhodium l tetrafluoroborate | |
(1Z,5Z)-cycloocta-1,5-diene;(2S,5S)-1-[2-[(2S,5S)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane;rhodium;tetrafluoroborate |
RUO â Research Use Only