Please login to your online account to display your discounted pricing

1,2-Bis((2R,5R)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, ACROS Organics™


Quantity 100mg

Manufacturer: thermo fisher scientific  AC435221000

 View more versions of this product

Catalog No. AC435221000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 556.21
Quantity 100mg
Color Orange to red to brown
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
CAS 305818-67-1
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Elemental Analysis 45.6 to 49.4% C
Chemical Name or Material 1,2-Bis((2R,5R)-2,5-Dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate
Infrared Spectrum Authentic
NMR (1H-NMR) Consistent with structure, (31P-NMR) Consistent with structure, NMR (19F-NMR), Consistent with structure
Molecular Formula C22H40P2Rh·BF4
MDL Number MFCD09265087

Specifications

Chemical Name or Material 1,2-Bis((2R,5R)-2,5-Dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate
CAS 305818-67-1
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C22H40P2Rh·BF4
Formula Weight 556.21
MDL Number MFCD09265087
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents