missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(-)-1,2-Bis[(2R,5R)-2,5-dimethylphospholano]ethane(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar 47064MC

 View more versions of this product

Catalog No. AA47064MC



Specifications

Specifications

(-)-1,2-Bis[(2R,5R)-2,5-dimethylphospholano]ethane(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate
Orange to Red
177750-25-3
MFCD01631282
LQQSHTWLDKZMBA-ZTCBNWEGSA-M
(1Z,5Z)-cycloocta-1,5-diene;(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]ethyl]-2,5-dimethylphospholane;rhodium;trifluoromethanesulfonate
5702643
Solid
Air Sensitive
100mg
C23H40F3O3P2RhS-
+-1,2-bis 2r,5r-2,5-dimethyl phospholano ethane cyclooctadiene rhodium i trifluoro methane sulfonate, +-1,2-bis-2r,5r-2,5-dimethylphospholano ethane cyclooctadiene rhodium i trifluoromethanesulfonate, 1z,5z-cycloocta-1,5-diene; 2r,5r-1-2-2r,5r-2,5-dimethylphospholan-1-yl ethyl-2,5-dimethylphospholane; rhodium; trifluoromethanesulfonate
CC1CCC(P1CCP2C(CCC2C)C)C.C1CC=CCCC=C1.C(F)(F)(F)S(=O)(=O)[O-].[Rh]
618.478
618.48
97%

Chemical Identifiers

177750-25-3
618.478
LQQSHTWLDKZMBA-ZTCBNWEGSA-M
5702643
CC1CCC(P1CCP2C(CCC2C)C)C.C1CC=CCCC=C1.C(F)(F)(F)S(=O)(=O)[O-].[Rh]
C23H40F3O3P2RhS-
MFCD01631282
+-1,2-bis 2r,5r-2,5-dimethyl phospholano ethane cyclooctadiene rhodium i trifluoro methane sulfonate, +-1,2-bis-2r,5r-2,5-dimethylphospholano ethane cyclooctadiene rhodium i trifluoromethanesulfonate, 1z,5z-cycloocta-1,5-diene; 2r,5r-1-2-2r,5r-2,5-dimethylphospholan-1-yl ethyl-2,5-dimethylphospholane; rhodium; trifluoromethanesulfonate
(1Z,5Z)-cycloocta-1,5-diene;(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]ethyl]-2,5-dimethylphospholane;rhodium;trifluoromethanesulfonate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents