Please login to your online account to display your discounted pricing

(-)-1,2-Bis[(2R,5R)-2,5-dimethylphospholano]ethane(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate, 97%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material (-)-1,2-Bis[(2R,5R)-2,5-dimethylphospholano]ethane(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate
CAS 177750-25-3
Molecular Formula C23H40F3O3P2RhS
Formula Weight 618.48
MDL Number MFCD01631282
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA47064MC

 
alfa aesar
47064MC
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA47064ME

 
alfa aesar
47064ME
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 618.48
Quantity 100mg
Chemical Name or Material (-)-1,2-Bis[(2R,5R)-2,5-dimethylphospholano]ethane(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate
Physical Form Solid
Assay 97%
Molecular Formula C23H40F3O3P2RhS
MDL Number MFCD01631282
CAS 177750-25-3
Color Orange to Red
Health Hazard 1 Warning
Sensitivity Air Sensitive
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.