missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]ethane, 97+%, Alfa Aesar™

$120.59 - $454.92

Chemical Identifiers

CAS 129648-07-3
Molecular Formula C14H28P2
Molecular Weight (g/mol) 258.326
MDL Number MFCD01073770
InChI Key IRCDUOCGSIGEAI-AAVRWANBSA-N
Synonym r,r-me-bpe, unii-1ls1afh1f1, 1ls1afh1f1, +-1,2-bis 2r,5r-2,5-dimethylphospholano ethane, s,s-me-en-duphos, me-bpe, r,r, +-1,2-bis 2r,5r-2,5-dimethylphospholano ethane, r,r-me-bpe mi, r,r-me-bpe, +, 2r,5r-1-2-2r,5r-2,5-dimethylphospholan-1-yl ethyl-2,5-dimethylphospholane
PubChem CID 2734555
IUPAC Name (2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]ethyl]-2,5-dimethylphospholane
SMILES CC1CCC(P1CCP2C(CCC2C)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60071MD View Documents Alfa Aesar
H60071MD
250mg Each for $120.59
 
AAH6007103 View Documents Alfa Aesar
H6007103
1g Each for $454.92
 
Specifications

Chemical Identifiers

129648-07-3
258.326
IRCDUOCGSIGEAI-AAVRWANBSA-N
2734555
CC1CCC(P1CCP2C(CCC2C)C)C
C14H28P2
MFCD01073770
r,r-me-bpe, unii-1ls1afh1f1, 1ls1afh1f1, +-1,2-bis 2r,5r-2,5-dimethylphospholano ethane, s,s-me-en-duphos, me-bpe, r,r, +-1,2-bis 2r,5r-2,5-dimethylphospholano ethane, r,r-me-bpe mi, r,r-me-bpe, +, 2r,5r-1-2-2r,5r-2,5-dimethylphospholan-1-yl ethyl-2,5-dimethylphospholane
(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]ethyl]-2,5-dimethylphospholane

Specifications

64°C to 67°C (0.06mmHg)
129648-07-3
MFCD01073770
r,r-me-bpe, unii-1ls1afh1f1, 1ls1afh1f1, +-1,2-bis 2r,5r-2,5-dimethylphospholano ethane, s,s-me-en-duphos, me-bpe, r,r, +-1,2-bis 2r,5r-2,5-dimethylphospholano ethane, r,r-me-bpe mi, r,r-me-bpe, +, 2r,5r-1-2-2r,5r-2,5-dimethylphospholan-1-yl ethyl-2,5-dimethylphospholane
CC1CCC(P1CCP2C(CCC2C)C)C
258.326
258.32
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]ethane
250mg
C14H28P2
UN2845
IRCDUOCGSIGEAI-AAVRWANBSA-N
(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]ethyl]-2,5-dimethylphospholane
2734555
≥97%
Safety and Handling

Safety and Handling

GHS H Statement
H250-H252-H315-H319-H335
Catches fire spontaneously if exposed to air.
Self-heating in large quantities; may catch fire.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P222-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not allow contact with air.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.