missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Bis((2R,5R)-2,5-diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC435200500

 View more versions of this product

Catalog No. AC435200500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1,2-Bis((2R,5R)-2,5-Diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate
Authentic
Brown to Yellow
569650-64-2
100.0
C34H56P2Rh·BF4
+-1,2-bis 2r,5r-2,5-di-i-propylphospholano benzene 1,5-cyclooctadiene rhodium i tetrafluoroborate, 1,2-bis 2r,5r-2,5-diisopropylphospholano benzene 1,5-cyclooctadiene rhodium i tetrafluoroborate, r,r-i-pr-duphos-rh
[B-](F)(F)(F)F.CC(C)C1CCC(P1C2=CC=CC=C2P3C(CCC3C(C)C)C(C)C)C(C)C.C1CC=CCCC=C1.[Rh]
716.48
716.48
97%
54.7 to 59.3% C
(1H-NMR) Consistent with structure, (31P-NMR) Consistent with structure
50mg
96.0
97%
MFCD07369039
IJKOQQHIAGRJHA-QPZXYOTCSA-N
(1Z,5Z)-cycloocta-1,5-diene;(2R,5R)-1-[2-[(2R,5R)-2,5-di(propan-2-yl)phospholan-1-yl]phenyl]-2,5-di(propan-2-yl)phospholane;rhodium;tetrafluoroborate
12964416
Powder

Chemical Identifiers

569650-64-2
716.48
IJKOQQHIAGRJHA-QPZXYOTCSA-N
12964416
[B-](F)(F)(F)F.CC(C)C1CCC(P1C2=CC=CC=C2P3C(CCC3C(C)C)C(C)C)C(C)C.C1CC=CCCC=C1.[Rh]
C34H56P2Rh·BF4
MFCD07369039
+-1,2-bis 2r,5r-2,5-di-i-propylphospholano benzene 1,5-cyclooctadiene rhodium i tetrafluoroborate, 1,2-bis 2r,5r-2,5-diisopropylphospholano benzene 1,5-cyclooctadiene rhodium i tetrafluoroborate, r,r-i-pr-duphos-rh
(1Z,5Z)-cycloocta-1,5-diene;(2R,5R)-1-[2-[(2R,5R)-2,5-di(propan-2-yl)phospholan-1-yl]phenyl]-2,5-di(propan-2-yl)phospholane;rhodium;tetrafluoroborate
SDS Documents