missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]ethane, 96%, Alfa Aesar™

Manufacturer:  Alfa Aesar H60593MD

 View more versions of this product

Catalog No. AAH60593MD



Description

Description

Applications
It is a DuPhos and BPE ligands which are highly efficient privileged ligands.

Notes
Air sensitive. Store at room temperature.
Specifications

Specifications

1,2-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]ethane
250mg
C22H44P2
1,2-bis 2s,5s-2,5-diisopropylphospholano ethane, 1,2-bis 2r,5r-2,5-di-i-propylphospholano ethane, 1,2-bis 2r,5r-2,5-diisopropylphospholan-1-yl ethane, 1,1'-ethylenebis 2r,5r-2,5-diisopropyltetrahydro-1h-phosphole, 1,2-bis 2r,5r-2,5-di-i-propylphospholano ethane, min, 1,2-bis 2r,5r-2,5-diisopropylphospholano ethane, kanata purity, 1,2-bis 2s,5s-2,5-diisopropylphospholano ethane, kanata purity, 2r,5r-1-2-2r,5r-2,5-diisopropylphospholan-1-yl ethyl-2,5-diisopropylphospholane
CC(C)C1CCC(P1CCP2C(CCC2C(C)C)C(C)C)C(C)C
370.542
370.53
67°C to 72°C
136705-63-0
MFCD08705243
IRVIFEWWPYKALC-GXRSIYKFSA-N
(2R,5R)-1-[2-[(2R,5R)-2,5-di(propan-2-yl)phospholan-1-yl]ethyl]-2,5-di(propan-2-yl)phospholane
11707202
96%

Chemical Identifiers

136705-63-0
370.542
IRVIFEWWPYKALC-GXRSIYKFSA-N
11707202
CC(C)C1CCC(P1CCP2C(CCC2C(C)C)C(C)C)C(C)C
C22H44P2
MFCD08705243
1,2-bis 2s,5s-2,5-diisopropylphospholano ethane, 1,2-bis 2r,5r-2,5-di-i-propylphospholano ethane, 1,2-bis 2r,5r-2,5-diisopropylphospholan-1-yl ethane, 1,1'-ethylenebis 2r,5r-2,5-diisopropyltetrahydro-1h-phosphole, 1,2-bis 2r,5r-2,5-di-i-propylphospholano ethane, min, 1,2-bis 2r,5r-2,5-diisopropylphospholano ethane, kanata purity, 1,2-bis 2s,5s-2,5-diisopropylphospholano ethane, kanata purity, 2r,5r-1-2-2r,5r-2,5-diisopropylphospholan-1-yl ethyl-2,5-diisopropylphospholane
(2R,5R)-1-[2-[(2R,5R)-2,5-di(propan-2-yl)phospholan-1-yl]ethyl]-2,5-di(propan-2-yl)phospholane
Documents