missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]benzene, 97+%, Alfa Aesar™

$242.11 - $882.09

Chemical Identifiers

CAS 136705-65-2
Molecular Formula C26H44P2
Molecular Weight (g/mol) 418.586
MDL Number MFCD02684549
InChI Key RBVGOQHQBUPSGX-MOUTVQLLSA-N
Synonym r,r-i-pr-duphos, +-1,2-bis 2r,5r-2,5-diisopropylphospholano benzene, 1,2-bis 2r,5r-2,5-diisopropylphospholan-1-yl benzene, +-1,2-bis 2r,5r-2,5-di-i-propylphospholano benzene, +-1,2-bis 2r,5r-2,5-diisopropylphospholano benzene, kanata purity, 2r,5r-1-2-2r,5r-2,5-diisopropylphospholan-1-yl phenyl-2,5-diisopropylphospholane
PubChem CID 11327562
IUPAC Name (2R,5R)-1-[2-[(2R,5R)-2,5-di(propan-2-yl)phospholan-1-yl]phenyl]-2,5-di(propan-2-yl)phospholane
SMILES CC(C)C1CCC(P1C2=CC=CC=C2P3C(CCC3C(C)C)C(C)C)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60227MD View Documents Alfa Aesar
H60227MD
250mg Each for $242.11
 
AAH6022703 View Documents Alfa Aesar
H6022703
1g Each for $882.09
 
Description

Description

Applications
It is used as ligands like DuPhos and BPE ligands and are highly efficient privileged ligands.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications

Chemical Identifiers

136705-65-2
418.586
RBVGOQHQBUPSGX-MOUTVQLLSA-N
11327562
CC(C)C1CCC(P1C2=CC=CC=C2P3C(CCC3C(C)C)C(C)C)C(C)C
C26H44P2
MFCD02684549
r,r-i-pr-duphos, +-1,2-bis 2r,5r-2,5-diisopropylphospholano benzene, 1,2-bis 2r,5r-2,5-diisopropylphospholan-1-yl benzene, +-1,2-bis 2r,5r-2,5-di-i-propylphospholano benzene, +-1,2-bis 2r,5r-2,5-diisopropylphospholano benzene, kanata purity, 2r,5r-1-2-2r,5r-2,5-diisopropylphospholan-1-yl phenyl-2,5-diisopropylphospholane
(2R,5R)-1-[2-[(2R,5R)-2,5-di(propan-2-yl)phospholan-1-yl]phenyl]-2,5-di(propan-2-yl)phospholane

Specifications

>110°C (230°F)
250mg
C26H44P2
r,r-i-pr-duphos, +-1,2-bis 2r,5r-2,5-diisopropylphospholano benzene, 1,2-bis 2r,5r-2,5-diisopropylphospholan-1-yl benzene, +-1,2-bis 2r,5r-2,5-di-i-propylphospholano benzene, +-1,2-bis 2r,5r-2,5-diisopropylphospholano benzene, kanata purity, 2r,5r-1-2-2r,5r-2,5-diisopropylphospholan-1-yl phenyl-2,5-diisopropylphospholane
CC(C)C1CCC(P1C2=CC=CC=C2P3C(CCC3C(C)C)C(C)C)C(C)C
418.586
418.58
1,2-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]benzene
+103° (c=1 in Chloroform)
136705-65-2
MFCD02684549
RBVGOQHQBUPSGX-MOUTVQLLSA-N
(2R,5R)-1-[2-[(2R,5R)-2,5-di(propan-2-yl)phospholan-1-yl]phenyl]-2,5-di(propan-2-yl)phospholane
11327562
≥97%