missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Bis((2R,5R)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC435181000

 View more versions of this product

Catalog No. AC435181000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1,2-Bis((2R,5R)-2,5-Diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate
Authentic
Brown to Orange
228121-39-9
100.0
C30H48P2Rh·BF4
XIOPHSBHVOCZMM-QFNVKMGQSA-N
cyclooctane;(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane;rhodium;tetrafluoroborate
11549211
Powder
52.4 to 56.8% C
(1H-NMR) Consistent with structure, (31P-NMR) Consistent with structure, NMR (19F-NMR), Consistent with structure
100mg
96.0
97%
MFCD01862464
[B-](F)(F)(F)F.CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC.C1C[CH][CH]CC[CH][CH]1.[Rh]
660.37
660.37
97%

Chemical Identifiers

228121-39-9
660.37
XIOPHSBHVOCZMM-QFNVKMGQSA-N
cyclooctane;(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane;rhodium;tetrafluoroborate
C30H48P2Rh·BF4
MFCD01862464
11549211
[B-](F)(F)(F)F.CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC.C1C[CH][CH]CC[CH][CH]1.[Rh]
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw

GHS Signal Word: Warning

SDS Documents