missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2-Bis((2R,5R)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC435182500
Specifications
1,2-Bis((2R,5R)-2,5-Diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate | |
Authentic | |
Brown to Orange | |
228121-39-9 | |
100.0 | |
C30H48P2Rh·BF4 | |
XIOPHSBHVOCZMM-QFNVKMGQSA-N | |
cyclooctane;(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane;rhodium;tetrafluoroborate | |
11549211 | |
Powder |
52.4 to 56.8% C | |
(1H-NMR) Consistent with structure, (31P-NMR) Consistent with structure, NMR (19F-NMR), Consistent with structure | |
250mg | |
96.0 | |
97% | |
MFCD01862464 | |
[B-](F)(F)(F)F.CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC.C1C[CH][CH]CC[CH][CH]1.[Rh] | |
660.37 | |
660.37 | |
97% |
Chemical Identifiers
228121-39-9 | |
660.37 | |
XIOPHSBHVOCZMM-QFNVKMGQSA-N | |
cyclooctane;(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane;rhodium;tetrafluoroborate |
C30H48P2Rh·BF4 | |
MFCD01862464 | |
11549211 | |
[B-](F)(F)(F)F.CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC.C1C[CH][CH]CC[CH][CH]1.[Rh] |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw
GHS Signal Word: Warning