Please login to your online account to display your discounted pricing

(-)-1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (-)-1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene
CAS 136705-64-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C22 H36 P2
Formula Weight 362.48
MDL Number MFCD00142335
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC363721000

  SDS
thermo fisher scientific
363721000
Glass bottle 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid or Low Melting Solid
Formula Weight 362.48
Quantity 100mg
Color Clear colorless
Merck Index 15, 3513
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material (-)-1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene
Assay Percent Range 97%
Molecular Formula C22 H36 P2
CAS 136705-64-1
Infrared Spectrum Authentic
Packaging Glass bottle
MDL Number MFCD00142335