missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(-)-1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 363721000

 View more versions of this product

Catalog No. AC363721000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
Liquid or Low Melting Solid
Clear colorless
96.0
136705-64-1
(-)-1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene
MFCD00142335
Authentic
362.48
100mg
15, 3513
100.0
C22 H36 P2
97%
Glass bottle

Chemical Identifiers

136705-64-1
100.0
C22 H36 P2
MFCD00142335
≥96.0%
96.0
97%
362.48
(-)-1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene