missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]ethane, 97+%, Alfa Aesar™

$198.93 - $653.86

Chemical Identifiers

CAS 136705-62-9
Molecular Formula C18H36P2
Molecular Weight (g/mol) 314.434
MDL Number MFCD01073768
InChI Key QOLRLVPABLMMKI-BRSBDYLESA-N
Synonym et-bpe, r,r, r,r-et-bpe, +-1,2-bis 2r,5r-2,5-diethylphospholano ethane, +-1,2-bis 2r,5r-2,5-diethylphospholano ethane, r,r-et-bpe mi, bpe ligands r,r-et-bpe mi, phospholane, 1,1'-1,2-ethanediyl bis 2,5-diethyl-, 2r,2'r,5r,5'r, +-1,2-bis 2r,5r-2,5-diethylphospholano ethane, 1,2-bis 2r,5r-2,5-diethylphospholan-1-yl ethane, 1,1'-ethylenebis 2alpha,5beta-diethyl-1-phosphacyclopentane
PubChem CID 11433485
IUPAC Name (2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]ethyl]-2,5-diethylphospholane
SMILES CCC1CCC(P1CCP2C(CCC2CC)CC)CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60447MD View Documents Alfa Aesar
H60447MD
250mg Each for $198.93
 
AAH6044703 View Documents Alfa Aesar
H6044703
1g Each for $653.86
 
Specifications

Chemical Identifiers

136705-62-9
314.434
QOLRLVPABLMMKI-BRSBDYLESA-N
11433485
CCC1CCC(P1CCP2C(CCC2CC)CC)CC
C18H36P2
MFCD01073768
et-bpe, r,r, r,r-et-bpe, +-1,2-bis 2r,5r-2,5-diethylphospholano ethane, +-1,2-bis 2r,5r-2,5-diethylphospholano ethane, r,r-et-bpe mi, bpe ligands r,r-et-bpe mi, phospholane, 1,1'-1,2-ethanediyl bis 2,5-diethyl-, 2r,2'r,5r,5'r, +-1,2-bis 2r,5r-2,5-diethylphospholano ethane, 1,2-bis 2r,5r-2,5-diethylphospholan-1-yl ethane, 1,1'-ethylenebis 2alpha,5beta-diethyl-1-phosphacyclopentane
(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]ethyl]-2,5-diethylphospholane

Specifications

110°C (230°F)
104°C to 106°C (0.05mmHg)
136705-62-9
MFCD01073768
et-bpe, r,r, r,r-et-bpe, +-1,2-bis 2r,5r-2,5-diethylphospholano ethane, +-1,2-bis 2r,5r-2,5-diethylphospholano ethane, r,r-et-bpe mi, bpe ligands r,r-et-bpe mi, phospholane, 1,1'-1,2-ethanediyl bis 2,5-diethyl-, 2r,2'r,5r,5'r, +-1,2-bis 2r,5r-2,5-diethylphospholano ethane, 1,2-bis 2r,5r-2,5-diethylphospholan-1-yl ethane, 1,1'-ethylenebis 2alpha,5beta-diethyl-1-phosphacyclopentane
CCC1CCC(P1CCP2C(CCC2CC)CC)CC
314.434
314.43
(+)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]ethane
+320° (c=1 in Hexane)
250mg
C18H36P2
UN2845
QOLRLVPABLMMKI-BRSBDYLESA-N
(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]ethyl]-2,5-diethylphospholane
11433485
≥97%
Safety and Handling

Safety and Handling

GHS H Statement
H250-H252-H315-H319-H335
Catches fire spontaneously if exposed to air.
Self-heating in large quantities; may catch fire.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P222-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not allow contact with air.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.